
Ouabain octahydrate

Synonym(s):
1β,3β,5β,11α,14,19-Hexahydroxycard-20(22)-enolide 3-(6-deoxy-α-L-mannopyranoside), Acocantherine, G-Strophanthin, Ouabain, Octahydrate
Empirical Formula (Hill Notation):
C29H44O12 · 8H2O
CAS Number:
Molecular Weight:
728.77
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing