O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol
Synonym(s):
α,ω-Bis-NHS-PEG, Di(N-succinimidyl) PEG-diacid, PEG-bis(N-succinimidyl succinate)
CAS Number:
Products
Brand
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing