Skip to Content
Merck
Octreotide ≥98% (HPLC)

Octreotide

Octreotide ≥98% (HPLC)
Synonym(s):
SMS 201-995
Empirical Formula (Hill Notation):
C49H66N10O10S2
CAS Number:
Molecular Weight:
1019.24

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing