Skip to Content
Merck
Ochratoxin A purum, ≥98.0% (TLC), from Aspergillus ochraceus

Ochratoxin A

Ochratoxin A purum, ≥98.0% (TLC), from Aspergillus ochraceus
Synonym(s):
N-[(3R)-(5-Chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl)carbonyl]-L-phenylalanine
Empirical Formula (Hill Notation):
C20H18ClNO6
CAS Number:
Molecular Weight:
403.81
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Mode of Action
Special Grade