N-tert-Butylmethylamine
Synonym(s):
N-Methyl-tert-butylamine
Linear Formula:
(CH3)3CNHCH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing