Nonanoic acid
Synonym(s):
Acid C9, Pelargonic acid, Nonanoic acid
Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6