Skip to Content
Merck
N-Nitrosodimethylamine (NDMA) Pharmaceutical Secondary Standard; Certified Reference Material

N-Nitrosodimethylamine (NDMA)

N-Nitrosodimethylamine (NDMA) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N-Methyl-N-nitrosomethanamine, N-Nitrosodimethylamine, Dimethylnitrosamine
Linear Formula:
(CH3)2NNO
CAS Number:
Molecular Weight:
74.08

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade