![N-Methyl-N-(trimethylsilyl)trifluoroacetamide for GC derivatization, LiChropur™, ≥98.5%](/deepweb/assets/sigmaaldrich/product/structures/350/565/8f6cad26-4deb-4bfd-95d9-6c112b2b4d07/640/8f6cad26-4deb-4bfd-95d9-6c112b2b4d07.png)
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
![N-Methyl-N-(trimethylsilyl)trifluoroacetamide for GC derivatization, LiChropur™, ≥98.5%](/deepweb/assets/sigmaaldrich/product/structures/350/565/8f6cad26-4deb-4bfd-95d9-6c112b2b4d07/640/8f6cad26-4deb-4bfd-95d9-6c112b2b4d07.png)
Synonym(s):
N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA, N-Methyl-N-trimethylsilyl-trifluoroacetamide
Linear Formula:
CF3CON(CH3)Si(CH3)3
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing