Skip to Content
Merck
N-Methyl-N-trimethylsilylheptafluorobutyramide for GC derivatization, LiChropur™, ≥90% (GC)

N-Methyl-N-trimethylsilylheptafluorobutyramide

N-Methyl-N-trimethylsilylheptafluorobutyramide for GC derivatization, LiChropur™, ≥90% (GC)
Synonym(s):
N-Trimethylsilyl-N-methylheptafluorobutyramide, MSHFBA
Linear Formula:
CF3CF2CF2CON(CH3)Si(CH3)3
CAS Number:
Molecular Weight:
299.26

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-1 of 1