Skip to Content
Merck
Nitrofurazone Related Compound A pharmaceutical secondary standard, certified reference material

Nitrofurazone Related Compound A

Nitrofurazone Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
5-Nitro-2-furfuraldazine
Empirical Formula (Hill Notation):
C10H6N4O6
CAS Number:
Molecular Weight:
278.18

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2