![Nilotinib Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/240/393/d59f3e62-9eb8-46cd-a48c-110d324c5799/640/d59f3e62-9eb8-46cd-a48c-110d324c5799.png)
Nilotinib Related Compound A
![Nilotinib Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/240/393/d59f3e62-9eb8-46cd-a48c-110d324c5799/640/d59f3e62-9eb8-46cd-a48c-110d324c5799.png)
Synonym(s):
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline
Empirical Formula (Hill Notation):
C11H10F3N3
CAS Number:
Molecular Weight:
241.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2