Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel - Nanocrystalline
Synonym(s):
Nickel - Nanocrystalline
Products
Manufacturer
Physical Form
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GF69920028
Nickel - Nanocrystalline,
foil, light tested, 12x12mm, thickness 0.01mm, 10-20nm nominal grain size, permanent copper support
Expand
View Pricing
GF55519083
Nickel - Nanocrystalline,
foil, light tested, 20x25mm, thickness 0.01mm, 10-20nm nominal grain size, permanent copper support
Expand
View Pricing
GF68381905
Nickel - Nanocrystalline,
foil, light tested, 25x25mm, thickness 0.05mm, 10-20nm nominal grain size, permanent copper support
Expand
View Pricing