![Nalorphine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/373/274/fd137b61-eb55-43dc-a41d-931ed5ae65eb/640/fd137b61-eb55-43dc-a41d-931ed5ae65eb.png)
Nalorphine hydrochloride
![Nalorphine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/373/274/fd137b61-eb55-43dc-a41d-931ed5ae65eb/640/fd137b61-eb55-43dc-a41d-931ed5ae65eb.png)
Synonym(s):
N-Allylnormorphine HCl, Nalline HCl
Empirical Formula (Hill Notation):
C19H21NO3 · HCl
CAS Number:
Molecular Weight:
347.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing