Skip to Content
Merck
N-Acetylcysteamine 95%

N-Acetylcysteamine

N-Acetylcysteamine 95%
Synonym(s):
N-(2-Mercaptoethyl)acetamide
Linear Formula:
CH3CONHCH2CH2SH
CAS Number:
Molecular Weight:
119.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing