N-(4-Phenylbenzylidene)benzylamine
Synonym(s):
N-(4-Biphenylylmethylene)benzylamine
Empirical Formula (Hill Notation):
C20H17N
CAS Number:
Molecular Weight:
271.36
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing