
N-(2-Aminoethyl)ethanolamine

Synonym(s):
N-(2-Hydroxyethyl)ethylenediamine, 2-(2-Aminoethylamino)ethanol, N-(2-Aminoethyl)ethanolamine
Linear Formula:
NH2CH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Products
Brand
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing