MOPS sodium salt
Synonym(s):
3-(N-Morpholino)propanesulfonic acid sodium salt, 4-Morpholinepropanesulfonic acid sodium salt, MOPS, Sodium, ULTROL® Grade, MOPS sodium salt
Empirical Formula (Hill Notation):
C7H14NNaO4S
CAS Number:
Molecular Weight:
231.25
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing