![Monobenzone pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/403/174/aa5237a1-16d6-4a68-b243-1f0be6bdc562/640/aa5237a1-16d6-4a68-b243-1f0be6bdc562.png)
Monobenzone
![Monobenzone pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/403/174/aa5237a1-16d6-4a68-b243-1f0be6bdc562/640/aa5237a1-16d6-4a68-b243-1f0be6bdc562.png)
Synonym(s):
4-(Benzyloxy)phenol, 4-(Phenylmethoxy)phenol, Hydroquinone monobenzyl ether
Linear Formula:
C6H5CH2OC6H4OH
CAS Number:
Molecular Weight:
200.23
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing