Skip to Content
Merck
Methyl cis-10-heptadecenoate ≥99% (GC), liquid

Methyl cis-10-heptadecenoate

Methyl cis-10-heptadecenoate ≥99% (GC), liquid
Synonym(s):
cis-10-Heptadecenoic acid methyl ester
Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing