Skip to Content
Merck
Methyl-13C,d3-amine hydrochloride 99 atom % D, 99 atom % 13C

Methyl-13C,d3-amine hydrochloride

Methyl-13C,d3-amine hydrochloride 99 atom % D, 99 atom % 13C
Linear Formula:
13CD3NH2·HCl
CAS Number:
Molecular Weight:
71.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing