Methoxypolyethylene glycol azide
Synonym(s):
α-Methoxy-ω-azido-PEG, O-(2-Azidoethyl)-O′-methylpolyethylene glycol, PEG-Azide, mPEG Azide
Linear Formula:
N3CH2CH2(OCH2CH2)nOCH3
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Functional Group
Reaction Type
Showing 1-4 of 4