Skip to Content
Merck
MES hydrate free-flowing, Redi-Dri™, ≥99.0%

MES hydrate

MES hydrate free-flowing, Redi-Dri™, ≥99.0%
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid hydrate, 4-Morpholineethanesulfonic acid
Empirical Formula (Hill Notation):
C6H13NO4S · xH2O
CAS Number:
Molecular Weight:
195.24 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-7 of 7