Skip to Content
Merck
vibrant-m

Mebendazole Related Compound D

vibrant-m
Synonym(s):
Methyl 5-benzoyl-1-methylbenzimidazol-2-ylcarbamate, 1-Methyl mebendazole
Empirical Formula (Hill Notation):
C17H15N3O3
CAS Number:
Molecular Weight:
309.32

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2