L-Tyrosine-15N
Synonym(s):
(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid-15N, 3-(4-Hydroxyphenyl)-L-alanine-15N, L-4-Hydroxyphenylalanine-15N, 15N Labeled L-tyrosine
Linear Formula:
4-(OH)C6H4CH2CH(15NH2)CO2H
CAS Number:
Molecular Weight:
182.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing