Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Phenyl-d
5
-alanine
Linear Formula:
C
6
D
5
CH
2
CH(NH
2
)CO
2
H
CAS Number:
56253-90-8
Molecular Weight:
170.22
Products
Product Category
amino acids (3)
stable isotopes (3)
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
GMP (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
615870
L-Phenyl-d
5
-alanine,
≥98 atom % D, ≥99% (CP)
Expand
View Pricing
661619
L-Phenyl-d
5
-alanine,
endotoxin tested, 98 atom % D
Expand
View Pricing
W6586
L-Phenyl-d
5
-alanine,
API for Clinical Studies, 98 atom % D
Expand
View Pricing