Skip to Content
Merck
L-Phenylalanine tert-butyl ester hydrochloride ≥99.0% (HPLC)

L-Phenylalanine tert-butyl ester hydrochloride

L-Phenylalanine tert-butyl ester hydrochloride ≥99.0% (HPLC)
Linear Formula:
C6H5CH2CH(NH2)COOC(CH3)3 · HCl
CAS Number:
Molecular Weight:
257.76
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing