Skip to Content
Merck
Lithium bis(trimethylsilyl)amide 99.9% trace metals basis

Lithium bis(trimethylsilyl)amide

Lithium bis(trimethylsilyl)amide 99.9% trace metals basis
Synonym(s):
Hexamethyldisilazane lithium salt
Linear Formula:
[(CH3)3Si]2NLi
CAS Number:
Molecular Weight:
167.33

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing