Listeria monocytogenes serovar 4b WDCM 00021 Vitroids
Products
Product Category
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing