
Listeria monocytogenes serovar 1/2a WDCM 00109 Vitroids™

Products
Product Category
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing