![Levofloxacin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/831/df75dfb6-5265-4bce-8224-8c6332672807/640/df75dfb6-5265-4bce-8224-8c6332672807.png)
Levofloxacin
![Levofloxacin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/831/df75dfb6-5265-4bce-8224-8c6332672807/640/df75dfb6-5265-4bce-8224-8c6332672807.png)
Synonym(s):
(−)-Ofloxacin
Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing