Skip to Content
Merck
Leucoside ≥90% (LC/MS-UV)

Leucoside

Leucoside ≥90% (LC/MS-UV)
Synonym(s):
Kaempferol 3-sambubioside
Empirical Formula (Hill Notation):
C26H28O15
CAS Number:
Molecular Weight:
580.49

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing