Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead-tin alloy, Pb75Sn25
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF84308238
Lead-tin alloy, Pb75Sn25,
10m wire, 0.9mm diameter, as drawn
Expand
View Pricing
GF37946852
Lead-tin alloy, Pb75Sn25,
10m wire, 1mm diameter, as drawn
Expand
View Pricing
GF05937981
Lead-tin alloy, Pb75Sn25,
5m wire, 1mm diameter, as drawn
Expand
View Pricing
GF23971125
Lead-tin alloy, Pb75Sn25,
5m wire, 0.9mm diameter, as drawn
Expand
View Pricing
GF16532591
Lead-tin alloy, Pb75Sn25,
100m wire, 1mm diameter, as drawn
Expand
View Pricing
GF45599754
Lead-tin alloy, Pb75Sn25,
100m wire, 0.9mm diameter, as drawn
Expand
View Pricing
GF81644788
Lead-tin alloy, Pb75Sn25,
20m wire, 1mm diameter, as drawn
Expand
View Pricing
GF99439936
Lead-tin alloy, Pb75Sn25,
50m wire, 0.9mm diameter, as drawn
Expand
View Pricing
GF42220156
Lead-tin alloy, Pb75Sn25,
20m wire, 0.9mm diameter, as drawn
Expand
View Pricing
GF21806546
Lead-tin alloy, Pb75Sn25,
50m wire, 1mm diameter, as drawn
Expand
View Pricing