Skip to Content
Merck
L-Aspartic acid-13C4,15N 98 atom % 13C, 98 atom % 15N, 95% (CP)

L-Aspartic acid-13C4,15N

L-Aspartic acid-13C4,15N 98 atom % 13C, 98 atom % 15N, 95% (CP)
Linear Formula:
HO213C13CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
138.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing