L-Alanine
Synonym(s):
(S)-2-Aminopropionic acid, L-α-Aminopropionic acid, L-Alanine
Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
pH Value
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing