
L-Alaninamide hydrochloride

Linear Formula:
CH3CH(NH2)CONH2·HCl
CAS Number:
Molecular Weight:
124.57
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1