
Lactose Monohydrate

Synonym(s):
α-Lactose monohydrate, β-D-Gal-(1→4)-α-D-Glc, 4-O-β-D-Galactopyranosyl-α-D-glucose, Milk sugar
Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Products
Product Category
Brand
Formula Weight
Format
Analyte Suitability
Application
Optical Isomer
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing