Lacosamide impurity F
Synonym(s):
2-Acetamido-N-benzyl-3-hydroxypropionamide
Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing