Skip to Content
Merck
L-4-Thiazolidinecarboxylic acid 98%

L-4-Thiazolidinecarboxylic acid

L-4-Thiazolidinecarboxylic acid 98%
Synonym(s):
L-Thiaproline
Empirical Formula (Hill Notation):
C4H7NO2S
CAS Number:
Molecular Weight:
133.17
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing