![Ketorolac Tromethamine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/416/924/0357d688-34c6-4d0b-8915-056b14532947/640/0357d688-34c6-4d0b-8915-056b14532947.png)
Ketorolac Tromethamine
![Ketorolac Tromethamine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/416/924/0357d688-34c6-4d0b-8915-056b14532947/640/0357d688-34c6-4d0b-8915-056b14532947.png)
Synonym(s):
Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol
Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2