Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isohexane
Synonym(s):
Isohexane
Empirical Formula (Hill Notation):
C
6
H
14
CAS Number:
92112-69-1
Molecular Weight:
86.18
Products
Product Category
solvents (3)
Brand
Supelco (3)
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.04333
Isohexane,
≥95.0% (C₆H₁₄-isomeres, GC) or analysis EMSURE
®
Expand
View Pricing
1.04335
Isohexane,
≥99.0% (GC), suitable for HPLC
Expand
View Pricing
1.04340
Isohexane,
≥99.8% (GC), suitable for gas chromatography (GC) ECD and FID, SupraSolv
®
Expand
View Pricing