Skip to Content
Merck
Iron(II) sulfate hydrate 99.999% trace metals basis

Iron(II) sulfate hydrate

Iron(II) sulfate hydrate 99.999% trace metals basis
Synonym(s):
Sulfuric acid, iron(II) salt (1:1), hydrate
Linear Formula:
FeSO4 · xH2O
CAS Number:
Molecular Weight:
151.91 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
pH Value
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade