Irinotecan Related Compound A
Synonym(s):
(S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (S)-10-Hydroxycamptothecin
Empirical Formula (Hill Notation):
C20H16N2O5
CAS Number:
Molecular Weight:
364.35
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2