Ipratropium bromide Related Compound A
Synonym(s):
(1R,3r,5S,8r)-3-Hydroxy-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane bromide, N-Isopropyltropinium bromide
Empirical Formula (Hill Notation):
C11BrH22NO
CAS Number:
Molecular Weight:
264.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2