Skip to Content
Merck
Indole-3-butyric acid LAB

Indole-3-butyric acid

Indole-3-butyric acid LAB
Synonym(s):
4-(3-Indolyl)butanoic acid, 4-(3-Indolyl)butyric acid, IBA, Indole-3-butyric acid
Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3