IL-12 human
Synonym(s):
CTL maturation factor (TCMF),, Cytotoxic lymphocyte maturation factor (CLMF), NKSF, TSF, IL-12
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Recombinant Host
Quality Segment
Technique
Showing 1-2 of 2