Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
IKA
®
C-MAG HS digital IKAMAG
™
hot plate magnetic stirrers
Synonym(s):
hot plate magnetic stirrers, hot plate stirrers, magnetic stirrers, stirrers
Products
Manufacturer
Capacity (L)
Diameter (mm)
Length (mm)
Plate Material
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
Z767700
IKA
®
C-MAG HS digital IKAMAG
™
hot plate magnetic stirrers,
model, HS 4, AC/DC input 230 V AC, Schuko plug
Expand
View Pricing
Z767778
IKA
®
C-MAG HS digital IKAMAG
™
hot plate magnetic stirrers,
model, HS 10, AC/DC input 230 V AC, Schuko plug
Expand
View Pricing