Skip to Content
Merck
ICBA 99% (HPLC)

ICBA

ICBA 99% (HPLC)
Synonym(s):
1′,1′′,4′,4′′-Tetrahydro-di[1,4]methanonaphthaleno[1,2:2′,3′,56,60:2′′,3′′][5,6]fullerene-C60, C60 derivative, indene-C60 bisadduct
Empirical Formula (Hill Notation):
C78H16
CAS Number:
Molecular Weight:
952.96

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing