Skip to Content
Merck
Ibutilide fumarate United States Pharmacopeia (USP) Reference Standard, monograph mol wt. 885.23 ([(C20H36N2O3S)2 . C4H4O4])

Ibutilide fumarate

Ibutilide fumarate United States Pharmacopeia (USP) Reference Standard, monograph mol wt. 885.23 ([(C20H36N2O3S)2 . C4H4O4])
Synonym(s):
Ibutilide hemifumarate salt, (±)-N-[4-[4-(Ethylheptylamino)-1-hydroxybutyl]phenyl]-methanesulfonamide hemifumarate salt, Corvert
Empirical Formula (Hill Notation):
C20H36N2O3S · 0.5C4H4O4
CAS Number:
Molecular Weight:
442.61

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade