Skip to Content
Merck
Hydroquinonesulfonic acid potassium salt technical grade

Hydroquinonesulfonic acid potassium salt

Hydroquinonesulfonic acid potassium salt technical grade
Synonym(s):
2,5-Dihydroxybenzenesulfonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulfonate
Linear Formula:
(HO)2C6H3SO3K
CAS Number:
Molecular Weight:
228.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing