Skip to Content
Merck
Hydrogen chloride – 2-propanol solution ~1.25 M HCl (T), for GC derivatization, LiChropur™

Hydrogen chloride solution

Hydrogen chloride – 2-propanol solution ~1.25 M HCl (T), for GC derivatization, LiChropur™
Synonym(s):
Hydrochloric acid in 2-propanol
Linear Formula:
HCl · CH3CH(OH)CH3
Molecular Weight:
36.46

Products

Brand
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1